Learn More
Medchemexpress LLC HY-P1806A 1mg Medchemexpress, Gastrin I (1-14), human (TFA) CAS: Purity:>98%
SDP
Catalog No. 501934247
Encompass_Preferred
Medchemexpress, HY-P1806A 1mg Gastrin I (1-14), human (TFA) CAS: Gastrin I (1-14), human TFA is 1-14 fragment of human gastrin I peptide. Gastrin I is an endogenous, gastrointestinal peptide hormone. Gastrin is the major hormonal regulator of gastric acid secretion [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4247 Supplier Medchemexpress LLC Supplier No. HYP1806A1MG
Add to Cart
Add to Cart
Medchemexpress, HY-P1806A 1mg Gastrin I (1-14), human (TFA) CAS: Gastrin I (1-14), human TFA is 1-14 fragment of human gastrin I peptide. Gastrin I is an endogenous, gastrointestinal peptide hormone. Gastrin is the major hormonal regulator of gastric acid secretion [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.