Learn More
Selleck Chemical LLC Gastrin I human P1249-1MG
SDP
Catalog No. 5000040246
Encompass_Preferred
Gastrin I human is the endogenous peptide produced in the stomach and increases gastric acid secretion via cholecystokinin 2 (CCK2) receptor
Catalog No. 50-000-40246 Supplier Selleck Chemical LLC Supplier No. P12491MG
Gastrin I human is the endogenous peptide produced in the stomach and increases gastric acid secretion via cholecystokinin 2 (CCK2) receptor