Learn More
Bachem Gastrin I (human)
SDP
Supplier:  Bachem 4030646.0001
Encompass_Preferred
Gastrin I (human), Pyr-Gly-Pro-Trp-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Met-Asp-Phe-NH2, 10047-33-3, C97H124N20O31S, Mr 2098.23, 1mg. Little Gastrin (human), Gastrin-17 (human), Gastrin 1, human.
Catalog No. 50-194-6919