Learn More
Enzo Life Sciences Gastrin I (rat), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900149
Encompass_Preferred
The Gastrin I (rat), EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. Ready-to-use liquid color-coded reagents are provided to reduce error in the lab. This kit utilizes low sample volume and the ability to measure up to 37 samples in duplicate per 96-well plate. Sensitivity: 78.1 pg/ml (range 78.1 - 5,000 pg/ml). UniProt ID: P04563. Assay Time: 5 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Rat . Content: GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.
Catalog No. 50-200-6854