Learn More
CPC Scientific H-Val-Pro-Leu-Pro-Ala-Gly-Gly-Gly-Thr-Val-Leu-Thr-Lys-Met-Tyr-Pro-Arg-Gly-Asn-His-Trp-Ala-Val-Gly-His-Leu-Met-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific GAST003B
Encompass
Peptide that stimulates pancreatic and gastric acid secretion.ONE-LETTER SEQUENCE: VPLPAGGGTVLTKMYPRGNHWAVGHLM-NH2MOLECULAR FORMULA: C130H204N38O31S2MOLECULAR WEIGHT:2859.4STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [93755-85-2], RESEARCH AREA: GastrointestinalREFERENCES: E.R. Spindel et al., PNAS, 81, 5699 (1984)
Catalog No. 50-210-9320