Learn More
Cayman Chemical Parishin B, Purity: ≥98%, CAS: 174972-79-3, 10 mg
SDP
Catalog No. 502912354
Encompass_Preferred
A phenolic glycoside with anti-asthmatic activity; decreases specific airway resistance and leukocyte infiltration in BALF in a guinea pig model of allergen-induced asthma stimulated by ovalbumin inhalation at 50 mg/kg
Catalog No. 50-291-2354 Supplier Cayman Chemical Supplier No. 3629610 MG
A phenolic glycoside with anti-asthmatic activity; decreases specific airway resistance and leukocyte infiltration in BALF in a guinea pig model of allergen-induced asthma stimulated by ovalbumin inhalation at 50 mg/kg