Learn More
Apexbio Technology LLC Gastrodenol 57644-54-9 500mg
SDP
Catalog No. 5000090160
Encompass_Preferred
Gastrodenol is an orally active agent exerting antibacterial activity primarily through inhibition of Helicobacter pylori (H pylori) growth It is designed to disrupt H pylori colonization thereby contributing to reduced gastrointestinal inflammation and facilitating mucosal defense and repair Gastrodenol (Bismuth potassium citrate) exerts its biological activity primarily through inhibition of H pylori Preclinical studies indicate that particularly in combination with proton pump inhibitors such as Omeprazole it enhances targeted control of H pylori Based on these pharmacological properties Gastrodenol holds research potential in gastrointestinal pathology including peptic ulceration gastric malignancies and mechanisms related to tubulointerstitial nephritis
Catalog No. 50-000-90160 Supplier Apexbio Technology LLC Supplier No. C7152500
Add to Cart
Add to Cart
Gastrodenol is an orally active agent exerting antibacterial activity primarily through inhibition of Helicobacter pylori (H pylori) growth It is designed to disrupt H pylori colonization thereby contributing to reduced gastrointestinal inflammation and facilitating mucosal defense and repair Gastrodenol (Bismuth potassium citrate) exerts its biological activity primarily through inhibition of H pylori Preclinical studies indicate that particularly in combination with proton pump inhibitors such as Omeprazole it enhances targeted control of H pylori Based on these pharmacological properties Gastrodenol holds research potential in gastrointestinal pathology including peptic ulceration gastric malignancies and mechanisms related to tubulointerstitial nephritis