Learn More
Cayman Chemical Gastrodin, Purity: ≥98%, CAS: 62499-27-8, 100 mg
SDP
Catalog No. 502941065
Encompass_Preferred
A phenolic glycoside with diverse biological activities; reduces Aβ42-induced cell death of rat hippocampal neurons and BV-2 mouse microglia; improves learning and memory in various transgenic mouse models of Alzheimer's disease; reduces motor deficits and loss of dopaminergic neurons in a rat model of Parkinson's disease; reduces seizure duration and intensity in a rat model of pentylenetetrazole-kindling chronic epileptic seizures from 50-200 mg/kg; increases social interaction and decreases marble burying and grooming behaviors, as well as restores inhibitory synaptic transmission and GAT1 and α5 subunit-containing GABAA expression in the basolateral amygdala in a mouse model of valproic acid-induced autistic-like phenotypes at 100 mg/kg; increases biomechanical bone strength in a rat model of dexamethasone-induced osteoporosis; inhibits ethanol-induced hepatocellular apoptosis and restores liver function in a rat model of alcoholic liver disease
Catalog No. 50-294-1065 Supplier Cayman Chemical Supplier No. 28828100 MG
A phenolic glycoside with diverse biological activities; reduces Aβ42-induced cell death of rat hippocampal neurons and BV-2 mouse microglia; improves learning and memory in various transgenic mouse models of Alzheimer's disease; reduces motor deficits and loss of dopaminergic neurons in a rat model of Parkinson's disease; reduces seizure duration and intensity in a rat model of pentylenetetrazole-kindling chronic epileptic seizures from 50-200 mg/kg; increases social interaction and decreases marble burying and grooming behaviors, as well as restores inhibitory synaptic transmission and GAT1 and α5 subunit-containing GABAA expression in the basolateral amygdala in a mouse model of valproic acid-induced autistic-like phenotypes at 100 mg/kg; increases biomechanical bone strength in a rat model of dexamethasone-induced osteoporosis; inhibits ethanol-induced hepatocellular apoptosis and restores liver function in a rat model of alcoholic liver disease