Learn More
Biomatik Corporation Gastrodin (GTD) ELISA Kit
Supplier:  Biomatik Corporation EKU10861
Encompass_Preferred
Gastrodin (GTD) ELISA Kit for detecting GTD in biological agents, other biological fluids. Species Reactivity: Universal. Detection Range: 246.9-20000ng/ml. Sensitivity: 87.5ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Gastrodine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5883