Learn More
Aviva Systems Biology Corp GATA1 Antibody - N-terminal region (P100834_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100834_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GATA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GATA1. Alternate names: ERYF1, GF1, NFE1, GF-1, XLTT, GATA-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-62735