Learn More
Aviva Systems Biology Corp GATA2 Antibody - N-terminal region (ARP31855_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31855_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GATA2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human GATA2. Alternate names: DCML, NFE1B, MONOMACApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54341