Learn More
Aviva Systems Biology Corp GATA5 Antibody - C-terminal region (ARP36860_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36860_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GATA5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of mouse GATA5. Alternate names: ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-014-56286