Learn More
Aviva Systems Biology Corp GATA5 Antibody - middle region (ARP31627_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31627_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GATA5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GATA5. Alternate names: bB379O24.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Sheep
Catalog No. 50-014-53936