Learn More
Aviva Systems Biology Corp GATA6 Antibody - N-terminal region (ARP32550_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32550_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GATA6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GATA6. Alternate names: -ApplicationIF, WB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-54650