Learn More
Aviva Systems Biology Corp Gatad2b Antibody - C-terminal region (ARP39430_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39430_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gatad2b. It was validated on Western Blot. . Alternate names: AL118180, C430014D17Rik, KIAA1150, P66beta, mKIAA1150ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-805