Learn More
Electron Microscopy Sciences Gatan 3View SEM Pin Stubs Large, 2.4 mm Head, 2 x 12.5 mm Pin
SDP
Catalog No. NC1386387
Encompass
The Gatan 3View systems use special sample pins. These are used inside SEMs to create 3D imaging data of samples by serial slicing and BSD imaging. These pins are fully compatible with the Gatan 3View systems; 2 mm pin, 3 mm head with cone top and total height of 12.5 mm. Manufactured from vacuum grade aluminium.
Head: 2.4 mm
Pin: 2 x 12.5 mm
Catalog No. NC1386387 Supplier Electron Microscopy Sciences Supplier No. 7595903
The Gatan 3View systems use special sample pins. These are used inside SEMs to create 3D imaging data of samples by serial slicing and BSD imaging. These pins are fully compatible with the Gatan 3View systems; 2 mm pin, 3 mm head with cone top and total height of 12.5 mm. Manufactured from vacuum grade aluminium.
Head: 2.4 mm
Pin: 2 x 12.5 mm