Learn More
Aviva Systems Biology Corp GATC Antibody - N-terminal region (ARP63251_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63251_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GATC. It was validated on Western Blot. . Alternate names: 15E1.2, FLJ37000, MGC129938ApplicationWB. Species reactivity: Cow, Human, Pig, Rabbit
Catalog No. 50-287-922