Learn More
Perkin Elmer US LLC Gateway Controller for ATR Trough Top Plate - ZnSe, UK
Catalog No. 502113265
Encompass_Preferred
The controller has a temperature range from ambient to 200°C, in 1°C increments. This controller is compatible with the Spectrum 3/Frontier spectrometer.
Catalog No. 50-211-3265 Supplier Perkin Elmer US LLC Supplier No. L1272484
Add to Cart
Add to Cart
Request a Quote
The controller has a temperature range from ambient to 200°C, in 1°C increments. This controller is compatible with the Spectrum 3/Frontier spectrometer.