Learn More
Gateway Safety Coveralls™ Safety Glasses
Catalog No. 19037401
Click to view available options
No. per Pack:
10/Pk.
Catalog No. No. per Pack
19-037-401 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 19-037-401 Supplier Gateway Safety Supplier No. 6880
This item is not returnable. View return policy
Specifications
Certifications/Compliance Meets ANSI Z87 and CSA Z94.3 standards
Lens Tint Clear
No. per Pack 10/Pk.
Lens Material Polycarbonate