Learn More
MSC Gateway Safety Safety Glasses, Scratch-Resistant, Polycarbonate, Clear Lens, 1 ea
Catalog No. 5000802815
Encompass
Gateway Safety StarLite safety glasses offer exceptional eye protection with an original, lightweight design. These glasses provide all-around impact protection, making them ideal for various industrial and laboratory environments.
  • Incredibly lightweight, weighing less than one ounce
  • Deep, universal-fit temples provide excellent side protection
  • Polycarbonate lens material offers high impact resistance
  • Scratch-resistant lenses ensure clear vision and durability
  • Frameless, wrap-around design for enhanced field of view
  • Provides UV protection
Catalog No. 50-008-02815 Supplier MSC Supplier No. 09003575
Gateway Safety StarLite safety glasses offer exceptional eye protection with an original, lightweight design. These glasses provide all-around impact protection, making them ideal for various industrial and laboratory environments.
  • Incredibly lightweight, weighing less than one ounce
  • Deep, universal-fit temples provide excellent side protection
  • Polycarbonate lens material offers high impact resistance
  • Scratch-resistant lenses ensure clear vision and durability
  • Frameless, wrap-around design for enhanced field of view
  • Provides UV protection