Learn More
Selleck Chemical LLC Gatifloxacin hydrochloride
SDP
Catalog No. 5000003321
Encompass_Preferred
Gatifloxacin (AM-1155 CG5501 BMS-206584 PD135432) hydrochloride is a potent fluoroquinolone antibiotic with broad-spectrum antibacterial activity Gatifloxacin hydrochloride inhibits bacterial type II topoisomerases with IC50 of 13 8 g/ml and 0 109 g/ml for S aureus topoisomerase IV and E coli DNA gyrase respectively
Catalog No. 50-000-03321 Supplier Selleck Chemical LLC Supplier No. S596425MG
Gatifloxacin (AM-1155 CG5501 BMS-206584 PD135432) hydrochloride is a potent fluoroquinolone antibiotic with broad-spectrum antibacterial activity Gatifloxacin hydrochloride inhibits bacterial type II topoisomerases with IC50 of 13 8 g/ml and 0 109 g/ml for S aureus topoisomerase IV and E coli DNA gyrase respectively