Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Gatifloxacin sesquihydrate

Click to view available options
Quantity:
1g
5g
25g
112811-59-3
C19H22FN3O4
375.4
MFCD22373645
XUBOMFCQGDBHNK-UHFFFAOYSA-N
5379
CHEBI:5280
1-cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid
CC1CN(CCN1)C2=C(C=C3C(=C2OC)N(C=C(C3=O)C(=O)O)C4CC4)F
112811-59-3
C19H22FN3O4
375.4
MFCD22373645
XUBOMFCQGDBHNK-UHFFFAOYSA-N
5379
CHEBI:5280
1-cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid
CC1CN(CCN1)C2=C(C=C3C(=C2OC)N(C=C(C3=O)C(=O)O)C4CC4)F

A broad spectrum antibiotic

Merck Index 14,4376
Solubility Information Soluble in 1N NaOH at 10mg/mL
Formula Weight 402.42
Physical Form Powder
Boiling Point 684°C
Color White to Yellow
Quantity 5g
Chemical Name or Material Gatifloxacin sesquihydrate

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : VB1983600