Learn More
STEMCELL Technologies Gatifloxacin, Size: 1 g
SDP
Supplier:  STEMCELL Technologies 72752
Encompass
Gatifloxacin is a fluoroquinolone antibiotic which inhibits bacterial DNA gyrase (IC₅₀ = 0.109 µg/ml) and topoisomerase IV (IC₅₀ = 13.8 µg/ml; Takei et al.). It is much less effective against HeLa cell topoisomerase II (IC₅₀ = 265 µg/ml; Takei et al.).MAINTENANCE AND SELF-RENEWAL· Promotes self-renewal in cultured human and mouse embryonic stem cells (Desbordes et al.).
Catalog No. 50-197-5973