Learn More
MSC Perma-Patch Asphalt Patch, 5 gal, Black, Fast-Setting
Catalog No. 502835822
Encompass
Perma-Patch Gator-Aid is an asphaltic emulsion designed for repairing alligator cracks and revitalizing worn asphalt surfaces. Suitable for driveways, parking lots, or athletic courts, it offers professional-grade results. This high-performance mastic filler is specifically formulated for cracks wider than 1 inch and alligatored areas.
  • Fast-setting formula
  • Professional-grade durability
  • Repairs alligator cracks
  • Revitalizes worn asphalt surfaces
  • Suitable for cracks wider than 1 inch
  • Ideal for driveways, parking lots, or athletic courts
  • No blistering or re-emulsification
  • 0 VOC content
Catalog No. 50-283-5822 Supplier MSC Supplier No. 72194418
Perma-Patch Gator-Aid is an asphaltic emulsion designed for repairing alligator cracks and revitalizing worn asphalt surfaces. Suitable for driveways, parking lots, or athletic courts, it offers professional-grade results. This high-performance mastic filler is specifically formulated for cracks wider than 1 inch and alligatored areas.
  • Fast-setting formula
  • Professional-grade durability
  • Repairs alligator cracks
  • Revitalizes worn asphalt surfaces
  • Suitable for cracks wider than 1 inch
  • Ideal for driveways, parking lots, or athletic courts
  • No blistering or re-emulsification
  • 0 VOC content