Learn More
Cole-Parmer Wika 213.53 2.5 0 to 100 psi Industrial Gauge with Bottom Connection, 2 1/2" Dial
Catalog No. 50993694 Shop All Cole-Parmer Products
Encompass_Preferred
Gauges feature a copper alloy tube, brass socket, 304 stainless steel case, and plastic lens. All connections are 1/4" NPT(M). Shpg wt 1 lb (0.5 kg).
Catalog No. 50-993-694 Supplier Cole-Parmer Supplier No. 6804612
Gauges feature a copper alloy tube, brass socket, 304 stainless steel case, and plastic lens. All connections are 1/4" NPT(M). Shpg wt 1 lb (0.5 kg).