Learn More
Cole-Parmer Ashcroft 10843.0 3" Test Pressure Gauge 0-200 psi, 1/4" NPT Lower
Catalog No. 50992685 Shop All Cole-Parmer Products
Encompass_Preferred
Ashcroft Test Gauge, 0.5% Accuracy, 316 SS Bourdon Tube, 3 in. Dial, 0 to 100 psi, 0 to 200 psi
Catalog No. 50-992-685 Supplier Cole-Parmer Supplier No. 6803407
Ashcroft Test Gauge, 0.5% Accuracy, 316 SS Bourdon Tube, 3 in. Dial, 0 to 100 psi, 0 to 200 psi