Learn More
Cole-Parmer Ashcroft 10843.0 Test Gauge 0.5% Accuracy, 316 SS Bourdon Tube, 3" Dial, 0 to 30 psi
Catalog No. 50992682 Shop All Cole-Parmer Products
Encompass_Preferred
Ashcroft Test Gauge, 0.5% Accuracy, 316 SS Bourdon Tube, 3 in. Dial, 0 to 30 psi
Catalog No. 50-992-682 Supplier Cole-Parmer Supplier No. 6803404
Ashcroft Test Gauge, 0.5% Accuracy, 316 SS Bourdon Tube, 3 in. Dial, 0 to 30 psi