Learn More
Cole-Parmer Ashcroft 10824.5 4.5" Test Pressure Gauge 0-30 psi, 1/4" NPT Lower
Catalog No. 50992673 Shop All Cole-Parmer Products
Encompass_Preferred
Gauges are factory calibrated to ensure accuracy. To determine the right gauge for your application, choose the Bourdon tube and process connection material according to the conditions and medium to which the gauge will be exposed. Then choose your desired range and accuracy. For added safety, the maximum continuous pressure applied to the gauge should not exceed 75% of the maximum pressure of the gauge. All gauges have a 1/4'' NPT(M) process connection. The 3'' gauges include a soft carrying case; order hard carrying case 68030-90 for 4-1/2'' and 6'' dial gauges.
Catalog No. 50-992-673 Supplier Cole-Parmer Supplier No. 6803004
Gauges are factory calibrated to ensure accuracy. To determine the right gauge for your application, choose the Bourdon tube and process connection material according to the conditions and medium to which the gauge will be exposed. Then choose your desired range and accuracy. For added safety, the maximum continuous pressure applied to the gauge should not exceed 75% of the maximum pressure of the gauge. All gauges have a 1/4'' NPT(M) process connection. The 3'' gauges include a soft carrying case; order hard carrying case 68030-90 for 4-1/2'' and 6'' dial gauges.