Learn More
Cole-Parmer Wika 111.12 1.5 1.5" Standard Pressure Gauge, 0 to 160 psi, Back Mount
Catalog No. 50992209 Shop All Cole-Parmer Products
Encompass_Preferred
Our Standard Pressure Gauges are designed for long and reliable service under rugged conditions. Some typical applications are for pumps, hydraulic and pneumatic systems, compressors, and many more applications where liquid and gases do not corrode brass. The accuracy of the gauges is ± 3/2/3% of span in all sizes and they are available from 30 psi to 5000 psi. Vacuum ranges and compound ranges are available upon request.
Catalog No. 50-992-209 Supplier Cole-Parmer Supplier No. 6843326
Add to Cart
Add to Cart
Our Standard Pressure Gauges are designed for long and reliable service under rugged conditions. Some typical applications are for pumps, hydraulic and pneumatic systems, compressors, and many more applications where liquid and gases do not corrode brass. The accuracy of the gauges is ± 3/2/3% of span in all sizes and they are available from 30 psi to 5000 psi. Vacuum ranges and compound ranges are available upon request.