Learn More
Cole-Parmer Wika 213.53 2.5 2.5" Filled Industrial Pressure Gauge, 0 to 10000 psi, Lower
Catalog No. 50992169 Shop All Cole-Parmer Products
Encompass_Preferred
These gauges feature a stainless steel case for protection in harsh environments. These gauges are supplied standard with a glycerine filled case. Suitable for environments compatible with copper alloy wetted parts where vibration or pressure pulsation occur and for gaseous or liquid media that will not obstruct the pressure system.
Catalog No. 50-992-169 Supplier Cole-Parmer Supplier No. 6843270
These gauges feature a stainless steel case for protection in harsh environments. These gauges are supplied standard with a glycerine filled case. Suitable for environments compatible with copper alloy wetted parts where vibration or pressure pulsation occur and for gaseous or liquid media that will not obstruct the pressure system.