Learn More
Cole-Parmer Wika 232.54 2.5 Industrial Stainless Steel Gauge, 0 to 30 psi
Catalog No. 50992159 Shop All Cole-Parmer Products
Encompass_Preferred
These gauges feature 316 stainless steel wetted parts and 304 stainless steel case and bayonet ring, an adjustable pointer, a laminated safety glass window, and can be liquid-filled in the field. WIKA stainless steel gauges are ideal for process, chemical, and petrochemical applications, oil exploration and production, power generation, pollution control equipment, and any other application requiring fine quality, long lasting pressure measurement instrumentation.
  • Center Back Mount Connection Available
  • 1/4" NPT Connection Size
  • 4" Dial Size Available
Catalog No. 50-992-159 Supplier Cole-Parmer Supplier No. 6843257
These gauges feature 316 stainless steel wetted parts and 304 stainless steel case and bayonet ring, an adjustable pointer, a laminated safety glass window, and can be liquid-filled in the field. WIKA stainless steel gauges are ideal for process, chemical, and petrochemical applications, oil exploration and production, power generation, pollution control equipment, and any other application requiring fine quality, long lasting pressure measurement instrumentation.
  • Center Back Mount Connection Available
  • 1/4" NPT Connection Size
  • 4" Dial Size Available