Learn More
Cole-Parmer Wika 9768343 GAUGE 2.5IN 30PSI 1/4 B
Supplier:  Cole-Parmer 6843438
Encompass_Preferred
These all stainless steel gauges are designed to meet API and industry-specific specifications. Rugged construction makes these pressure gauges well suited for tough process industries including chemical, petroleum/refining, power stations, onshore and offshore mining, environmental technology, and new plant construction.
Catalog No. 50-992-253