Learn More
Cole-Parmer Wika 111.10 2.5 2.5" Standard Pressure Gauge, 0 to 200 psi, Lower Mount
Catalog No. 50992206 Shop All Cole-Parmer Products
Encompass_Preferred
Our standard pressure gauges are designed for long and reliable service under rugged conditions. Typical applications are for pumps, hydraulic and pneumatic systems, compressors, as a contractor's gauge and for many other applications where the measured media does not corrode brass.
Catalog No. 50-992-206 Supplier Cole-Parmer Supplier No. 6843323
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Our standard pressure gauges are designed for long and reliable service under rugged conditions. Typical applications are for pumps, hydraulic and pneumatic systems, compressors, as a contractor's gauge and for many other applications where the measured media does not corrode brass.