Learn More
Cole-Parmer Wika 213.40 Forged Brass Industrial Gauge ~ 0 To 1000 PSI, 2 1/2"
Catalog No. 50992183 Shop All Cole-Parmer Products
Encompass_Preferred
These high-quality gauges are intended for use in adverse service conditions where pulsation or vibration exists. The solid forged-brass case and socket is resistant to shock and vibration, and is suitable for hydraulics and compressors. Liquid fill is glycerine.
Catalog No. 50-992-183 Supplier Cole-Parmer Supplier No. 6843285
These high-quality gauges are intended for use in adverse service conditions where pulsation or vibration exists. The solid forged-brass case and socket is resistant to shock and vibration, and is suitable for hydraulics and compressors. Liquid fill is glycerine.