Learn More
Cole-Parmer Wika 9318178 GAUGE 2.5IN 100PSI 1/4 B
Supplier:  Cole-Parmer 6843472
Encompass_Preferred
These high-quality gauges are intended for use in adverse service conditions where pulsation or vibration exists. The solid forged-brass case and socket is resistant to shock and vibration, and is suitable for hydraulics and compressors. Liquid fill is glycerine.
Catalog No. 50-992-277