Learn More
Cole-Parmer Wika 213.40 2.5 Forged Brass Industrial Gauge, 2.5" dial, 1/4"NPT(M), 0 to 1500 psi
Catalog No. 50992229 Shop All Cole-Parmer Products
Encompass_Preferred
These high-quality gauges are intended for use in adverse service conditions where pulsation or vibration exists. The solid forged-brass case and socket is resistant to shock and vibration, and is suitable for hydraulics and compressors. Liquid fill is glycerine.
Catalog No. 50-992-229 Supplier Cole-Parmer Supplier No. 6843373
These high-quality gauges are intended for use in adverse service conditions where pulsation or vibration exists. The solid forged-brass case and socket is resistant to shock and vibration, and is suitable for hydraulics and compressors. Liquid fill is glycerine.