Learn More
Cole-Parmer Wika 9318275 GAUGE 2.5IN 3000PSI 1/4 B
Catalog No. 50992240 Shop All Cole-Parmer Products
Encompass_Preferred
These high-quality gauges are intended for use in adverse service conditions where pulsation or vibration exists. The solid forged-brass case and socket is resistant to shock and vibration, and is suitable for hydraulics and compressors. Liquid fill is glycerine.
Catalog No. 50-992-240 Supplier Cole-Parmer Supplier No. 6843393
These high-quality gauges are intended for use in adverse service conditions where pulsation or vibration exists. The solid forged-brass case and socket is resistant to shock and vibration, and is suitable for hydraulics and compressors. Liquid fill is glycerine.