Learn More
Cole-Parmer Wika 9851933 GAUGE 2.5IN 5PSI 1/4 L
Supplier:  Cole-Parmer 6843455
Encompass_Preferred
Our WIKA low pressure gauges are extremely sensitive and highly accurate. The copper alloy capsule is designed to measure pressure and vacuum of gaseous media from as low as 15" H2O to 240" H2O (10 psi). An adjustment screw is located on the dial to re-zero the pointer if necessary. The finely polished nickel silver pinion gear and shaft of the movement ensure repeatable accuracy.
Catalog No. 50-992-264