Learn More
Cole-Parmer Wika 611.10 2.5 2.5" Dual Scale Vacuum Gauge, 60" WC and 1500 mm H2O to 0
Catalog No. 50992187 Shop All Cole-Parmer Products
Encompass_Preferred
Our WIKA low pressure gauges are extremely sensitive and highly accurate. The copper alloy capsule is designed to measure pressure and vacuum of gaseous media from as low as 15" H2O to 240" H2O (10 psi). An adjustment screw is located on the dial to re-zero the pointer if necessary. The finely polished nickel silver pinion gear and shaft of the movement ensure repeatable accuracy.
Catalog No. 50-992-187 Supplier Cole-Parmer Supplier No. 6843289
Add to Cart
Add to Cart
Our WIKA low pressure gauges are extremely sensitive and highly accurate. The copper alloy capsule is designed to measure pressure and vacuum of gaseous media from as low as 15" H2O to 240" H2O (10 psi). An adjustment screw is located on the dial to re-zero the pointer if necessary. The finely polished nickel silver pinion gear and shaft of the movement ensure repeatable accuracy.