Learn More
Cole-Parmer Wika 111.10 2 2" Commercial Pressure Gauge, 0 to 30 psi, Lower Mount
Catalog No. 50992194 Shop All Cole-Parmer Products
Encompass_Preferred
Our standard pressure gauges are designed for long and reliable service under rugged conditions. Typical applications are for pumps, hydraulic and pneumatic systems, compressors, as a contractor's gauge and for many other applications where the measured media does not corrode brass.
Catalog No. 50-992-194 Supplier Cole-Parmer Supplier No. 6843306
Our standard pressure gauges are designed for long and reliable service under rugged conditions. Typical applications are for pumps, hydraulic and pneumatic systems, compressors, as a contractor's gauge and for many other applications where the measured media does not corrode brass.