Learn More
Cole-Parmer Wika 111.25 4.5 0 to 100 psi Industrial Gauge, 4 1/2" Dial
Catalog No. 50993705 Shop All Cole-Parmer Products
Encompass_Preferred
Gauges feature copper alloy tube, brass socket, 304 stainless steel case, and plastic lens. Bottom connection is 1/4" NPT(M). Shpg wt 1 lb (0.5 kg).
Catalog No. 50-993-705 Supplier Cole-Parmer Supplier No. 6804716
Gauges feature copper alloy tube, brass socket, 304 stainless steel case, and plastic lens. Bottom connection is 1/4" NPT(M). Shpg wt 1 lb (0.5 kg).