Learn More
Cole-Parmer Wika 232.34 4.5 4.5" Filled Process Pressure Gauge, 0 to 300 psi, Lower
Catalog No. 50992148 Shop All Cole-Parmer Products
Encompass_Preferred
Specifically designed for the chemical and petroleum processing industries, WIKA process gauges have the construction, materials, and engineering it takes to stand up to the toughest conditions around. These gauges are equipped with stainless steel wetted parts. It is engineered to deliver years of accurate service while with-standing vibration, corrosive media and corrosive environments, and a wide range of temperatures.
Catalog No. 50-992-148 Supplier Cole-Parmer Supplier No. 6843241
Specifically designed for the chemical and petroleum processing industries, WIKA process gauges have the construction, materials, and engineering it takes to stand up to the toughest conditions around. These gauges are equipped with stainless steel wetted parts. It is engineered to deliver years of accurate service while with-standing vibration, corrosive media and corrosive environments, and a wide range of temperatures.