Learn More
Cole-Parmer Wika 212.34 4.5 4.5" Process Vacuum Gauge, 30 to 0" Hg, Lower
Catalog No. 50992140 Shop All Cole-Parmer Products
Encompass_Preferred
Brass wetted parts
Specifically designed for the chemical and petroleum processing industries, WIKA 4-1/2" process gauges have the construction, materials, and engineering it takes to stand up to the toughest conditions around. These gauges are equipped with brass-wetted parts. It is engineered to deliver years of accurate service while withstanding vibration and a wide range of temperatures.
Catalog No. 50-992-140 Supplier Cole-Parmer Supplier No. 6843230
Brass wetted parts
Specifically designed for the chemical and petroleum processing industries, WIKA 4-1/2" process gauges have the construction, materials, and engineering it takes to stand up to the toughest conditions around. These gauges are equipped with brass-wetted parts. It is engineered to deliver years of accurate service while withstanding vibration and a wide range of temperatures.