Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Gauge 4 In Dia Low Side Blue 400 psi

Catalog No. 502757488
Encompass_Preferred

Gauge 4 In Dia Low Side Blue 400 psi

Catalog No. 50-275-7488 Supplier Grainger Supplier No. 14X856
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gauge 4 In Dia Low Side Blue 400 psi

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.