Learn More
Cole-Parmer Wika 9699142 GAUGE 4IN 300PSI 1/4 L
Supplier:  Cole-Parmer 6843505
Encompass_Preferred
These gauges feature a stainless steel case for protection in harsh environments. These gauges are supplied standard with a glycerine filled case. Suitable for environments compatible with copper alloy wetted parts where vibration or pressure pulsation occur and for gaseous or liquid media that will not obstruct the pressure system.
Catalog No. 50-992-296