Learn More
Cole-Parmer Wika 9314652 GAUGE 4IN 30PSI 1/4 L
Catalog No. 50992231 Shop All Cole-Parmer Products
Encompass_Preferred
These high-quality gauges are intended for use in adverse service conditions where pulsation or vibration exists. The solid forged-brass case and socket is resistant to shock and vibration, and is suitable for hydraulics and compressors. Liquid fill is glycerine.
Catalog No. 50-992-231 Supplier Cole-Parmer Supplier No. 6843380
These high-quality gauges are intended for use in adverse service conditions where pulsation or vibration exists. The solid forged-brass case and socket is resistant to shock and vibration, and is suitable for hydraulics and compressors. Liquid fill is glycerine.