Learn More
Cole-Parmer Ashcroft 14902.5 Dual-Scale Vacuum Gauge 100" WC and 25 kPa
Catalog No. 50992735 Shop All Cole-Parmer Products
Encompass_Preferred
Gauges have brass socket, beryllium copper diaphragm capsule, painted steel case, and polycarbonate lens. Bottom connection is 1/4" NPT(M).
Catalog No. 50-992-735 Supplier Cole-Parmer Supplier No. 6893033
Gauges have brass socket, beryllium copper diaphragm capsule, painted steel case, and polycarbonate lens. Bottom connection is 1/4" NPT(M).