Learn More
Med Vet International McKesson Blood Pressure Gauge, 01-802GM, Each
Catalog No. 503045573
Encompass_Preferred
Blood Pressure Gauge McKesson Brand entrust™ Performance Plus Gauge. Black body, black face with white printing. For use with Deluxe Aneroid Sphygmomanometers (01-720 Series). 300mmHg, no-pin stop. Individually tested to conform to ANSI SP9 standards. Not made with natural rubber latex. Packaged: Each.
Catalog No. 50-304-5573 Supplier Med Vet International Supplier No. 363782EA
Blood Pressure Gauge McKesson Brand entrust™ Performance Plus Gauge. Black body, black face with white printing. For use with Deluxe Aneroid Sphygmomanometers (01-720 Series). 300mmHg, no-pin stop. Individually tested to conform to ANSI SP9 standards. Not made with natural rubber latex. Packaged: Each.