Learn More
Med Vet International McKesson Blood Pressure Unit Gauge, 01-800GM, Each
Catalog No. 503045566
Encompass_Preferred
Blood Pressure Gauge McKesson Brand Chrome-plated body, black face with white printing For use with Professional Aneroid Sphygmomanometers (01-700 Series). 300mmHg, no-pin stop. Individually tested to conform to ANSI SP9 standards. Not made with natural rubber latex. Packaged: Each
Catalog No. 50-304-5566 Supplier Med Vet International Supplier No. 363781EA
Blood Pressure Gauge McKesson Brand Chrome-plated body, black face with white printing For use with Professional Aneroid Sphygmomanometers (01-700 Series). 300mmHg, no-pin stop. Individually tested to conform to ANSI SP9 standards. Not made with natural rubber latex. Packaged: Each