Learn More
Cole-Parmer Shimpo FGE-100XY Compact gauge, 100 lb capacity
Supplier:  Cole-Parmer 9395409
Encompass
Digital force gauges feature 180° displays for readable results from any angle. Other features include dual-direction keypad, selectable force direction (push/pull), and selectable power on/off settings. Ergonomically designed gauges fit comfortably in your hand. Rugged all-metal construction gauges are available in a variety of capacities to fit the needs of your application. Universal power operation with included AC adapter/charger (100 to 240 VAC).
Catalog No. 50-993-605